accessibility menu, dialog, popup

UserName

CAS RN 25462-23-1

H3COHOH

CAS RN 25462-23-1

IUPAC Name: 2-pentylpropane-1,3-diol
Synonyms: 1,3-propanediol, 2-pentyl-
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: RHYUFGNCUXTFTC-UHFFFAOYSA-N
SMILES: CCCCCC(CO)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results