missing translation for 'onlineSavingsMsg'
Learn More

2-n-Octyl-4-isothiazolin-3-one 98.0+%, TCI America™

Catalog No. O03781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03781G Supplier TCI America Supplier No. O03781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26530-20-1
Molecular Formula C11H19NOS
Molecular Weight (g/mol) 213.339
MDL Number MFCD00072473
InChI Key JPMIIZHYYWMHDT-UHFFFAOYSA-N
PubChem CID 33528
ChEBI CHEBI:81936
IUPAC Name 2-octyl-1,2-thiazol-3-one
SMILES CCCCCCCCN1C(=O)C=CS1

Specifications

Boiling Point 142°C
Quantity 1 g
UN Number 2923
Formula Weight 213.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-n-Octyl-4-isothiazolin-3-one [for Biochemical Research]
EINECSNumber (5)-5246
RTECSNumber NX8156900
TSCA Yes