missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Octyl-1-dodecanol 90.0+%, TCI America™
$68.97 - $542.44
Chemical Identifiers
| CAS | 5333-42-6 |
|---|---|
| Molecular Formula | C20H42O |
| Molecular Weight (g/mol) | 298.56 |
| MDL Number | MFCD01310428 |
| InChI Key | LEACJMVNYZDSKR-UHFFFAOYNA-N |
| PubChem CID | 21414 |
| IUPAC Name | 2-octyldodecan-1-ol |
| SMILES | CCCCCCCCCCC(CO)CCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O042925ML
|
TCI America
O042925ML |
25 mL |
Each for $68.97
|
|
|||||
|
O0429500ML
|
TCI America
O0429500ML |
500 mL |
Each for $542.44
|
|
|||||
Chemical Identifiers
| 5333-42-6 | |
| 298.56 | |
| LEACJMVNYZDSKR-UHFFFAOYNA-N | |
| 2-octyldodecan-1-ol |
| C20H42O | |
| MFCD01310428 | |
| 21414 | |
| CCCCCCCCCCC(CO)CCCCCCCC |
Specifications
| 5333-42-6 | |
| Colorless | |
| C20H42O | |
| 25 mL | |
| CCCCCCCCCCC(CO)CCCCCCCC | |
| 298.56 | |
| 298.56 | |
| Liquid |
| 1°C | |
| 161°C | |
| MFCD01310428 | |
| LEACJMVNYZDSKR-UHFFFAOYNA-N | |
| 2-octyldodecan-1-ol | |
| 21414 | |
| ≥90.0% (GC) | |
| 2-n-Octyl-1-dodecanol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : JR4240000
TSCA : Yes