missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Octyl-1-dodecanol 90.0+%, TCI America™
Supplier: TCI America O0429500ML
Specifications
| 2-n-Octyl-1-dodecanol | |
| 1°C | |
| 161°C | |
| MFCD01310428 | |
| LEACJMVNYZDSKR-UHFFFAOYNA-N | |
| 2-octyldodecan-1-ol | |
| 21414 | |
| ≥90.0% (GC) |
| 5333-42-6 | |
| Colorless | |
| C20H42O | |
| 500 mL | |
| CCCCCCCCCCC(CO)CCCCCCCC | |
| 298.56 | |
| 298.56 | |
| Liquid |
Chemical Identifiers
| 5333-42-6 | |
| 298.56 | |
| LEACJMVNYZDSKR-UHFFFAOYNA-N | |
| 2-octyldodecan-1-ol |
| C20H42O | |
| MFCD01310428 | |
| 21414 | |
| CCCCCCCCCCC(CO)CCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : JR4240000
TSCA : Yes