missing translation for 'onlineSavingsMsg'
Learn More

2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAL1743306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1743306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1743306 Supplier Thermo Scientific Chemicals Supplier No. L1743306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 145100-50-1 | Molecular Formula: C7H4F6N2O4S2 | Molecular Weight: 358.23 g/mol

2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine is a reactive agent used for conversion of lithium enolates of ketones to vinyl triflates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine is a reactive agent used for conversion of lithium enolates of ketones to vinyl triflates.

Solubility
Hydrolyzes in water.

Notes
It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Store under inert gas.

Chemical Identifiers

CAS 145100-50-1
Molecular Formula C7H4F6N2O4S2
Molecular Weight (g/mol) 358.23
MDL Number MFCD00191834
InChI Key DXLQEJHUQKKSRB-UHFFFAOYSA-N
Synonym 2-n,n-bis trifluoromethylsulfonyl amino pyridine, 1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethyl sulfonyl methanesulfonamide, n-pyridin-2-yl triflimide, 2-n,n-bis trifluoromethyl sulphonylamino pyridine, n-2-pyridyl triflimide, n-2-pyridyl bis trifluoromethanesulfonimide, pyridin-2-amine, n,n-bis trifluoromethylsulfonyl, 2-bis trifluoromethylsulfonyl amino pyridine, 1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethanesulfonylmethanesulfonamide, n,n-bis trifluoromethylsulfonyl-2-pyridylamine
PubChem CID 534172
IUPAC Name 1,1,1-trifluoro-N-pyridin-2-yl-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES FC(F)(F)S(=O)(=O)N(C1=CC=CC=N1)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 37°C to 41°C
Boiling Point 97°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 5832565
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 358.24
Percent Purity 98%
Chemical Name or Material 2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only