missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-n-Hexylthiophene, 98%
Description
2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
Solubility
Soluble in most organic solvents and ethanol Soluble . Insoluble in water.
Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.
Solubility
Soluble in most organic solvents and ethanol Soluble . Insoluble in water.
Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Density | 0.935 |
| Boiling Point | 228°C to 230°C |
| Flash Point | 100°C (212°F) |
| Refractive Index | 1.496 |
| Quantity | 50 g |
| Beilstein | 110220 |
| Solubility Information | Soluble in most organic solvents and ethanol Soluble . Insoluble in water. |
| Formula Weight | 168.3 |
| Percent Purity | 98% |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| EINECSNumber | 242-579-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only