missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexylthiophene, 98%

Catalog No. AAA1265209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1265209 10 g
AAA1265218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1265209 Supplier Thermo Scientific Chemicals Supplier No. A1265209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18794-77-9 | C10H16S | 168.30 g/mol

2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in most organic solvents and ethanol Soluble . Insoluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 18794-77-9
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.30
MDL Number MFCD00022535
InChI Key QZVHYFUVMQIGGM-UHFFFAOYSA-N
Synonym thiophene, 2-hexyl, 2-n-hexylthiophene, 2-hexyl-thiophene, hexylthiophene, thiophene, hexyl, 2-hexyl thiophene, 2-? hexylthiophene, 2-hexylthiophene, ksc179k7d
PubChem CID 87793
IUPAC Name 2-hexylthiophene
SMILES CCCCCCC1=CC=CS1

Specifications

Density 0.935
Boiling Point 228°C to 230°C
Flash Point 100°C (212°F)
Refractive Index 1.496
Quantity 10 g
Beilstein 110220
Solubility Information Soluble in most organic solvents and ethanol Soluble . Insoluble in water.
Formula Weight 168.3
Percent Purity 98%
Chemical Name or Material 2-n-Hexylthiophene
EINECSNumber 242-579-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only