missing translation for 'onlineSavingsMsg'
Learn More

2-n-Hexyldecanoic acid, 97%

Catalog No. AAL1223822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1223822 100 g
AAL1223814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1223822 Supplier Thermo Scientific Chemicals Supplier No. L1223822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25354-97-6 | C16H32O2 | 256.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25354-97-6
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00059651
InChI Key JMOLZNNXZPAGBH-UHFFFAOYSA-N
Synonym decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16
PubChem CID 32912
IUPAC Name 2-hexyldecanoic acid
SMILES CCCCCCCCC(CCCCCC)C(=O)O

Specifications

Melting Point 10°C
Density 0.88
Boiling Point 165°C to 168°C (2 mmHg)
Flash Point 174°C (345°F)
Refractive Index 1.446
Quantity 100 g
Beilstein 1777217
Merck Index 14,4709
Formula Weight 256.43
Percent Purity 97%
Chemical Name or Material 2-n-Hexyldecanoic acid
Show More Show Less
EINECSNumber 246-885-9
RTECSNumber HD9750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only