Learn More
2-n-Butylfuran, 98%
CAS: 4466-24-4 | C8H12O | 124.183 g/mol
$1,482.86 - $1,482.86
Chemical Identifiers
| CAS | 4466-24-4 |
|---|---|
| Molecular Formula | C8H12O |
| Molecular Weight (g/mol) | 124.183 |
| MDL Number | MFCD00047071 |
| InChI Key | NWZIYQNUCXUJJJ-UHFFFAOYSA-N |
| Synonym | furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane |
| PubChem CID | 20534 |
| IUPAC Name | 2-butylfuran |
| SMILES | CCCCC1=CC=CO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1337630
|
Thermo Scientific Chemicals
A1337630 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4466-24-4 | |
| 124.183 | |
| NWZIYQNUCXUJJJ-UHFFFAOYSA-N | |
| 20534 | |
| CCCCC1=CC=CO1 |
| C8H12O | |
| MFCD00047071 | |
| furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane | |
| 2-butylfuran |
Specifications
| 4466-24-4 | |
| 153°C to 154°C | |
| Pleasant | |
| MFCD00047071 | |
| UN1993 | |
| furan, 2-butyl, 2-n-butyl furan, 2-n-butylfuran, unii-81jv9zyk0d, 81jv9zyk0d, 2-butyl furan, 2-butyl-furan, 2-but-1-yl fura, 2-but-1-yl furan, 1-fur-2-yl butane | |
| CCCCC1=CC=CO1 | |
| 124.183 | |
| 124.18 | |
| 2-n-Butylfuran |
| 0.886 | |
| 32°C (89°F) | |
| C8H12O | |
| 250 g | |
| 107045 | |
| NWZIYQNUCXUJJJ-UHFFFAOYSA-N | |
| 2-butylfuran | |
| 20534 | |
| 98% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 224-732-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only