missing translation for 'onlineSavingsMsg'
Learn More

2-(Morpholinothio)benzothiazole 97.0+%, TCI America™

Catalog No. M053225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M053225G 25 g
M0532500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M053225G Supplier TCI America Supplier No. M053225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102-77-2
Molecular Formula C11H12N2OS2
Molecular Weight (g/mol) 252.35
MDL Number MFCD00022870
InChI Key MHKLKWCYGIBEQF-UHFFFAOYSA-N
Synonym N-Oxydiethylene-2-benzothiazolesulfenamide
PubChem CID 7619
IUPAC Name 4-(1,3-benzothiazol-2-ylsulfanyl)morpholine
SMILES C1COCCN1SC2=NC3=CC=CC=C3S2

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 252.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Morpholinothio)benzothiazole
EINECSNumber (5)-0897
RTECSNumber DL5950000
TSCA Yes