missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinonicotinic acid, 97%, Thermo Scientific™

Catalog No. CC24601DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24601DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC24601DA Supplier Thermo Scientific Chemicals Supplier No. CC24601DA
Only null left

Chemical Identifiers

CAS 423768-54-1
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
MDL Number MFCD03086193
InChI Key YDUPMJIUJMCXAL-UHFFFAOYSA-N
Synonym 2-morpholinonicotinic acid, 2-morpholin-4-yl pyridine-3-carboxylic acid, 2-4-morpholino nicotinic acid, pubchem19585, 2-morpholinonicotinicacid, 2-morpholin-4-ylnicotinic acid, 2-morpholin-4-yl-nicotinic acid, 2-morpholinopyridine-3-carboxylic acid, 3-pyridinecarboxylicacid,2-4-morpholinyl
PubChem CID 2776568
IUPAC Name 2-morpholin-4-ylpyridine-3-carboxylic acid
SMILES C1COCCN1C2=C(C=CC=N2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.22
Percent Purity 97%
Chemical Name or Material 2-Morpholinonicotinic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.