missing translation for 'onlineSavingsMsg'
Learn More

[2-(Morpholinomethyl)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. CC48609DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC48609DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC48609DA Supplier Thermo Scientific Chemicals Supplier No. CC48609DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91271-63-5
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD03211251
InChI Key JOBIKMKNOFLXRQ-UHFFFAOYSA-N
Synonym 2-morpholinomethyl phenyl methanol, 2-morpholin-4-ylmethyl phenyl methanol, 2-morpholinomethyl-benzylalkohol, o-morpholin-1-yl methyl benzyl alcohol, benzenemethanol, 2-4-morpholinylmethyl, 2-morpholin-4-yl methyl phenyl methanol, 2-morpholin-4-ylmethyl phenyl methan-1-ol, 2-morpholin-4-yl-methyl-phenyl-methanol hydrochloride, 2-morpholin-4-ylmethyl phenyl methanol saltdata: hcl
PubChem CID 693121
IUPAC Name [2-(morpholin-4-ylmethyl)phenyl]methanol
SMILES C1COCCN1CC2=CC=CC=C2CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material [2-(Morpholinomethyl)phenyl]methanol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.