accessibility menu, dialog, popup

UserName

CAS RN 91271-63-5

IUPAC Name: {2-[(morpholin-4-yl)methyl]phenyl}methanol
Synonyms: [2-(morpholin-4-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: JOBIKMKNOFLXRQ-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)