missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinoethyl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. M29011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29011G 1 g
M29015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29011G Supplier TCI America Supplier No. M29011G
Only null left

Chemical Identifiers

CAS 2997-88-8
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00080577
InChI Key MNZNJOQNLFEAKG-UHFFFAOYSA-N
Synonym Methacrylic Acid 2-Morpholinoethyl Ester
PubChem CID 76343
IUPAC Name 2-morpholin-4-ylethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN1CCOCC1

Specifications

Color Red
Boiling Point 81°C
Quantity 1 g
Formula Weight 199.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Morpholinoethyl Methacrylate (stabilized with MEHQ)
EINECSNumber (5)-0866
TSCA No