missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one 98.0+%, TCI America™

Catalog No. M253710MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M253710MG 10 mg
M253750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M253710MG Supplier TCI America Supplier No. M253710MG

Chemical Identifiers

CAS 168425-64-7
Molecular Formula C16H15N3O2
Molecular Weight (g/mol) 281.315
MDL Number MFCD10568147
InChI Key BVRDQVRQVGRNHG-UHFFFAOYSA-N
PubChem CID 10039361
IUPAC Name 2-morpholin-4-ylpyrimido[2,1-a]isoquinolin-4-one
SMILES C1COCCN1C2=CC(=O)N3C=CC4=CC=CC=C4C3=N2

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 10 mg
Synonym compound 401, 2-morpholino-4h-pyrimido 2,1-a isoquinolin-4-one, 2-morpholin-4-yl pyrimido 2,1-a isoquinolin-4-one, 2-morpholin-4-yl-pyrimido 2,1-a isoquinolin-4-one, 2-4-morpholinyl-4h-pyrimido 2,1-a isoquinolin-4-one, compound401, d0t0by, 2-morpholin-1-yl pyrimido 2,1-a isoquinolin-4-one
Formula Weight 281.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one
TSCA No
Recommended Storage Refrigerator