missing translation for 'onlineSavingsMsg'
Learn More

2-Methylvaleric Acid 98.0+%, TCI America™

Catalog No. M061025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M061025ML 25 mL
M0610500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M061025ML Supplier TCI America Supplier No. M061025ML

Chemical Identifiers

CAS 97-61-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002671
InChI Key OVBFMEVBMNZIBR-UHFFFAOYSA-N
Synonym 2-methylvaleric acid, pentanoic acid, 2-methyl, valeric acid, 2-methyl, methylpropylacetic acid, 2-pentanecarboxylic acid, 2-methyl valeric acid, alpha-methylvaleric acid, pentanoic acid, methyl, kyselina 2-methylvalerova, fema no. 2754
PubChem CID 7341
IUPAC Name 2-methylpentanoic acid
SMILES CCCC(C)C(=O)O

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 mL
UN Number 3265
Formula Weight 116.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methylvaleric Acid
EINECSNumber (2)-0608
RTECSNumber YV7700000
TSCA Yes