missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiophene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018836
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018836 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018836 Supplier Merck Emd Millipore Supplier No. 80364100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Specifications

CAS 554-14-3
Product Line TraceCERT
Vapor Pressure 47.6 mmHg (37.7°C)
Density 1.014 g/mL (at 25°C)
Linear Formula C5H6S
MDL Number MFCD00005451
Melting Point -63°C
Boiling Point 113°C
Quantity 100 mg
Formula Weight 98.17
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC