missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiophene, 98%

CAS: 554-14-3 | C5H6S | 98.163 g/mol

$79.79 - $799.68

Chemical Identifiers

CAS 554-14-3
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005451
InChI Key XQQBUAPQHNYYRS-UHFFFAOYSA-N
Synonym thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
PubChem CID 11126
IUPAC Name 2-methylthiophene
SMILES CC1=CC=CS1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1273614
View Documents
Thermo Scientific Chemicals
A1273614
25 g
Each for $79.79
Only null left
 
AAA1273622
View Documents
Thermo Scientific Chemicals
A1273622
100 g
Each for $187.12
Only null left
 
AAA1273636
View Documents
Thermo Scientific Chemicals
A1273636
500 g
Each for $799.68
Only null left
 
Description

Description

2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases, strong oxidizing agents.
Specifications

Chemical Identifiers

554-14-3
98.163
XQQBUAPQHNYYRS-UHFFFAOYSA-N
11126
CC1=CC=CS1
C5H6S
MFCD00005451
thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
2-methylthiophene

Specifications

554-14-3
1.017
7°C (44°F)
C5H6S
MFCD00005451
UN1993
thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
XQQBUAPQHNYYRS-UHFFFAOYSA-N
2-methylthiophene
11126
98%
-63°C
112°C to 114°C
Aromatic
1.52
25 g
103734
Slightly soluble in water. Soluble in alcohol.
CC1=CC=CS1
98.163
98.17
2-Methylthiophene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 209-063-0

RTECSNumber : XM9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only