Learn More
2-Methylthiophene, 98%
CAS: 554-14-3 | C5H6S | 98.163 g/mol
Supplier: Thermo Scientific Chemicals A1273636
Description
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;
Solubility
Slightly soluble in water. Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases, strong oxidizing agents.
Specifications
| 2-Methylthiophene | |
| -63°C | |
| 112°C to 114°C | |
| Aromatic | |
| 1.52 | |
| 500 g | |
| 103734 | |
| Slightly soluble in water. Soluble in alcohol. | |
| CC1=CC=CS1 | |
| 98.163 | |
| 98.17 |
| 554-14-3 | |
| 1.017 | |
| 7°C (44°F) | |
| C5H6S | |
| MFCD00005451 | |
| UN1993 | |
| thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204 | |
| XQQBUAPQHNYYRS-UHFFFAOYSA-N | |
| 2-methylthiophene | |
| 11126 | |
| 98% |
Chemical Identifiers
| 554-14-3 | |
| 98.163 | |
| XQQBUAPQHNYYRS-UHFFFAOYSA-N | |
| 11126 | |
| CC1=CC=CS1 |
| C5H6S | |
| MFCD00005451 | |
| thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204 | |
| 2-methylthiophene |
Safety and Handling
GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 209-063-0
RTECSNumber : XM9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only