missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)thiophene, 97%

Catalog No. AAA1176314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1176314 25 g
AAA1176306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1176314 Supplier Thermo Scientific Chemicals Supplier No. A1176314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5780-36-9 | C5H6S2 | 130.223 g/mol

2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)thiophene is used as an intermediate in synthetic chemistry. It is also used in the preparation of 2-thiocyanato-thiophene by reaction with cyanogen iodide using sodium methylmercatide as a reagent.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5780-36-9
Molecular Formula C5H6S2
Molecular Weight (g/mol) 130.223
MDL Number MFCD00052382
InChI Key ZLSMPEVZXWDWEK-UHFFFAOYSA-N
PubChem CID 79844
IUPAC Name 2-methylsulfanylthiophene
SMILES CSC1=CC=CS1

Specifications

Boiling Point 82°C to 84°C (15 mmHg)
Odor Stench
Refractive Index 1.596
Quantity 25 g
UN Number UN3334
Beilstein 106272
Synonym 2-methylthio thiophene, thiophene, 2-methylthio, 2-methylsulfanyl thiophene, 2-methylthiothiophene, 2-mehtylthio thiophene, thiophene,2-methylthio, acmc-20aomi, 2-methylthio-thiophene, 2-methylmercaptothiophene, 2-methylsulfanyl thiophene #
Formula Weight 130.23
Percent Purity 97%
Chemical Name or Material 2- (Methylthio)thiophene
Hazard Category H319-H335
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only