missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)thiazole 98.0+%, TCI America™

Catalog No. p-7132818
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17125G 5 g
M171225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17125G Supplier TCI America Supplier No. M17125G
Only null left

Chemical Identifiers

CAS 5053-24-7
Molecular Formula C4H5NS2
Molecular Weight (g/mol) 131.21
MDL Number MFCD00858981
InChI Key VQNOAXZUEKPSJC-UHFFFAOYSA-N
Synonym 2-methylthio thiazole, 2-methylthiothiazole, 2-methylthio-1,3-thiazole, 2-methylsulfanyl-1,3-thiazole, thiazole, 2-methylthio, acmc-209knn, 2-methylsulfanyl-thiazole, thiazole,2-methylthio
PubChem CID 2760100
IUPAC Name 2-(methylsulfanyl)-1,3-thiazole
SMILES CSC1=NC=CS1

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 g
Formula Weight 131.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)thiazole