missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)pyrazine 98.0+%, TCI America™

Catalog No. M18215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18215G 5 g
M182125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18215G Supplier TCI America Supplier No. M18215G
Only null left

Chemical Identifiers

CAS 21948-70-9
Molecular Formula C5H6N2S
Molecular Weight (g/mol) 126.177
MDL Number MFCD00041024
InChI Key KBPBOWBQRUXMFV-UHFFFAOYSA-N
Synonym 2-methylthio pyrazine, 2-methylthiopyrazine, pyrazine, methylthio, pyrazinyl methyl sulfide, 2-methylsulfanyl pyrazine, methyl pyrazinyl sulfide, methylthio pyrazine, 2-methylsulfanyl-pyrazine, pyrazine, 2-methylthio, unii-r1c97e3a4f
PubChem CID 519907
IUPAC Name 2-methylsulfanylpyrazine
SMILES CSC1=NC=CN=C1

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 5 g
Formula Weight 126.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)pyrazine