missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M15701G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M15701G Supplier TCI America Supplier No. M15701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2-(Methylthio)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 168618-42-6
Melting Point 83°C
Molecular Formula C7H9BO2S
Quantity 1 g
MDL Number MFCD01318165
Synonym 2-methylthio phenylboronic acid, 2-methylthiophenylboronic acid, 2-methylthio phenyl boronic acid, 2-methylthio benzeneboronic acid, 2-thioanisoleboronic acid, 2-methylsulfanylphenyl boronic acid, 2-methylsulfanyl phenylboronic acid, 2-methylsulfanyl phenyl boranediol, 2-methylsulfanyl phenyl boronic acid, o-methylthio phenylboronic acid
InChI Key QXBWTYBCNFKURT-UHFFFAOYSA-N
SMILES CSC1=CC=CC=C1B(O)O
IUPAC Name [2-(methylsulfanyl)phenyl]boronic acid
Molecular Weight (g/mol) 168.02
PubChem CID 2773543
Formula Weight 168.02
Physical Form Crystalline Powder
Show More Show Less