missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)phenol 97.0+%, TCI America™

Catalog No. M18545G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18545G Supplier TCI America Supplier No. M18545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1073-29-6
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00002211
InChI Key SOOARYARZPXNAL-UHFFFAOYSA-N
Synonym 2-methylthio phenol, 2-hydroxythioanisole, 2-methylsulfanyl phenol, 2-hydroxy thioanisole, phenol, 2-methylthio, o-hydroxythioanisole, o-methylthio phenol, phenol, o-methylthio, 2-methylmercaptophenol, 2-methylmercapto phenol
PubChem CID 61261
IUPAC Name 2-(methylsulfanyl)phenol
SMILES CSC1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 140.20
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)phenol