missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethanol 98.0+%, TCI America™

Catalog No. p-7132119
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M035825ML 25 mL
M0358100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M035825ML Supplier TCI America Supplier No. M035825ML
Only null left

Chemical Identifiers

CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-(methylsulfanyl)ethan-1-ol
SMILES CSCCO

Specifications

Color Yellow
Boiling Point 81°C
Quantity 25 mL
Formula Weight 92.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)ethanol