missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethanol 98.0+%, TCI America™

Catalog No. m0358100ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
Catalog No. Quantity
M0358100ML 100 mL
M035825ML 25 mL
2 options

Catalog No. M0358100ML

Supplier: TCI America M0358100ML

Only null left

Chemical Identifiers

CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-(methylsulfanyl)ethan-1-ol
SMILES CSCCO

Specifications

Color Yellow
Boiling Point 81°C
Quantity 100 mL
Formula Weight 92.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)ethanol