missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethanol, 99%

Catalog No. p-3758493
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC210100050 5 mL Glass bottle
AC210100250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC210100050 Supplier Thermo Scientific Chemicals Supplier No. 210100050
Only null left

CAS: 5271-38-5 | C3H8OS | 92.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-methylsulfanylethanol
SMILES CSCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0600g/mL
Boiling Point 169°C to 171°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4934 to 1.4954
Linear Formula CH3SCH2CH2OH
Quantity 5 mL
Beilstein 01, II, 524
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble
Formula Weight 92.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)ethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-090-3

RUO – Research Use Only