missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethanol, 99%

Catalog No. aal0544214
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAL0544214 25 g
AAL0544206 5 g
AAL0544222 100 g
3 options

Catalog No. AAL0544214

Supplier: Thermo Scientific Chemicals L0544214

Only null left

CAS: 5271-38-5 | C3H8OS | 92.16 g/mol

2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible in water.

Notes
Do not store together with acids. Store away from Oxidizing agents.

Chemical Identifiers

CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-methylsulfanylethanol
SMILES CSCCO

Specifications

Density 1.06
Boiling Point 169°C to 171°C
Flash Point 70°C (158°F)
Odor Stench
Refractive Index 1.4944
Linear Formula CH3S(CH2)2OH
Quantity 25 g
UN Number UN3334
Beilstein 1731081
Solubility Information Not miscible in water.
Formula Weight 92.16
Percent Purity 99%
Chemical Name or Material 2-(Methylthio)ethanol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H302-H332-H227
Harmful if swallowed.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber 226-090-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only