missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethanol, 99%

Catalog No. AAL0544206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0544206 5 g
AAL0544214 25 g
AAL0544222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0544206 Supplier Thermo Scientific Chemicals Supplier No. L0544206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5271-38-5 | C3H8OS | 92.16 g/mol

2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible in water.

Notes
Do not store together with acids. Store away from Oxidizing agents.

Chemical Identifiers

CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-methylsulfanylethanol
SMILES CSCCO

Specifications

Density 1.06
Boiling Point 169°C to 171°C
Flash Point 70°C (158°F)
Odor Stench
Refractive Index 1.4944
Linear Formula CH3S(CH2)2OH
Quantity 5 g
UN Number UN3334
Beilstein 1731081
Solubility Information Not miscible in water.
Formula Weight 92.16
Percent Purity 99%
Chemical Name or Material 2-(Methylthio)ethanol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H302-H332-H227
Harmful if swallowed.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber 226-090-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only