missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylthio)benzothiazole 98.0+%, TCI America™
$115.57 - $894.75
Chemical Identifiers
| CAS | 615-22-5 |
|---|---|
| Molecular Formula | C8H7NS2 |
| Molecular Weight (g/mol) | 181.271 |
| MDL Number | MFCD00005784 |
| InChI Key | UTBVIMLZIRIFFR-UHFFFAOYSA-N |
| Synonym | 2-methylthio benzothiazole, 2-methylmercaptobenzothiazole, 2-methylthio benzo d thiazole, 2-methylthiobenzothiazole, 2-methylmercapto benzothiazole, benzothiazole, 2-methylthio, mtbt, 2-methylthio-1,3-benzothiazole, 2-methylsulfanyl-1,3-benzothiazole, 2-methyl mercaptobenzothiazole |
| PubChem CID | 11989 |
| ChEBI | CHEBI:1217 |
| IUPAC Name | 2-methylsulfanyl-1,3-benzothiazole |
| SMILES | CSC1=NC2=CC=CC=C2S1 |
Chemical Identifiers
| 615-22-5 | |
| 181.271 | |
| UTBVIMLZIRIFFR-UHFFFAOYSA-N | |
| 11989 | |
| 2-methylsulfanyl-1,3-benzothiazole |
| C8H7NS2 | |
| MFCD00005784 | |
| 2-methylthio benzothiazole, 2-methylmercaptobenzothiazole, 2-methylthio benzo d thiazole, 2-methylthiobenzothiazole, 2-methylmercapto benzothiazole, benzothiazole, 2-methylthio, mtbt, 2-methylthio-1,3-benzothiazole, 2-methylsulfanyl-1,3-benzothiazole, 2-methyl mercaptobenzothiazole | |
| CHEBI:1217 | |
| CSC1=NC2=CC=CC=C2S1 |
Specifications
| 615-22-5 | |
| White-Yellow | |
| C8H7NS2 | |
| 25 g | |
| UTBVIMLZIRIFFR-UHFFFAOYSA-N | |
| 2-methylsulfanyl-1,3-benzothiazole | |
| 11989 | |
| 181.27 | |
| Crystalline Powder |
| 46°C | |
| 177°C | |
| MFCD00005784 | |
| 2-methylthio benzothiazole, 2-methylmercaptobenzothiazole, 2-methylthio benzo d thiazole, 2-methylthiobenzothiazole, 2-methylmercapto benzothiazole, benzothiazole, 2-methylthio, mtbt, 2-methylthio-1,3-benzothiazole, 2-methylsulfanyl-1,3-benzothiazole, 2-methyl mercaptobenzothiazole | |
| CSC1=NC2=CC=CC=C2S1 | |
| 181.271 | |
| CHEBI:1217 | |
| ≥98.0% (GC) | |
| 2-(Methylthio)benzothiazole |
Safety and Handling
EINECSNumber : (5)-0244
TSCA : Yes