missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzonitrile, 98%

Catalog No. p-7044511
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0687003 1 g
AAL0687006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0687003 Supplier Thermo Scientific Chemicals Supplier No. L0687003

CAS: 6609-54-7 | C8H7NS | 149.211 g/mol

2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6609-54-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00015557
InChI Key PXZSANDJGNKIIA-UHFFFAOYSA-N
Synonym 2-methylthio benzonitrile, 2-methylsulfanyl benzonitrile, 2-cyanophenyl methyl sulfide, 2-methylmercapto benzonitrile, methylthio benzonitrile, 2-methylthiobenzenecarbonitrile, benzonitrile,2-methylthio, 2-methylsulfanyl benzenecarbonitrile
PubChem CID 138781
IUPAC Name 2-methylsulfanylbenzonitrile
SMILES CSC1=CC=CC=C1C#N

Specifications

Melting Point 33°C to 38°C
Boiling Point 116°C to 117°C (5 mmHg)
Quantity 1 g
UN Number UN3439
Beilstein 2207135
Solubility Information Insoluble in water.
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only