missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylthio)benzimidazole 99.0+%, TCI America™
Supplier: TCI America M043825G
Specifications
| 2-(Methylthio)benzimidazole | |
| 202°C | |
| C8H8N2S | |
| 25 g | |
| OCKJFOHZLXIAAT-UHFFFAOYSA-N | |
| 2-methylsulfanyl-1H-benzimidazole | |
| 23539 | |
| ≥99.0% (HPLC,T) |
| 7152-24-1 | |
| White | |
| MFCD00005594 | |
| 2-methylthio benzimidazole, 1h-benzimidazole, 2-methylthio, 2-methylmercaptobenzimidazole, 2-methylthio-1h-benzo d imidazole, 2-methylthiobenzimidazole, 2-methylsulfanyl-1h-benzimidazole, s-methyl-2-mercaptobenzimidazole, benzimidazole, 2-methylthio | |
| CSC1=NC2=CC=CC=C2N1 | |
| 164.226 | |
| 164.23 | |
| Crystalline Powder |
Chemical Identifiers
| 7152-24-1 | |
| 164.226 | |
| OCKJFOHZLXIAAT-UHFFFAOYSA-N | |
| 23539 | |
| CSC1=NC2=CC=CC=C2N1 |
| C8H8N2S | |
| MFCD00005594 | |
| 2-methylthio benzimidazole, 1h-benzimidazole, 2-methylthio, 2-methylmercaptobenzimidazole, 2-methylthio-1h-benzo d imidazole, 2-methylthiobenzimidazole, 2-methylsulfanyl-1h-benzimidazole, s-methyl-2-mercaptobenzimidazole, benzimidazole, 2-methylthio | |
| 2-methylsulfanyl-1H-benzimidazole |
Safety and Handling
EINECSNumber : (5)-0473
RTECSNumber : DD9480000
TSCA : Yes