missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)aniline 97.0+%, TCI America™

Catalog No. M115925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M115925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M115925G Supplier TCI America Supplier No. M115925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2987-53-3
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007708
InChI Key WBRPQQSADOCKCH-UHFFFAOYSA-N
Synonym 2-methylthio aniline, 2-aminothioanisole, 2-methylsulfanyl aniline, 2-methylmercaptoaniline, 2-methylmercapto aniline, o-thioanisidine, o-aminothioanisole, 2-amino thioanisole, benzenamine, 2-methylthio, o-methylthio aniline
PubChem CID 76337
IUPAC Name 2-methylsulfanylaniline
SMILES CSC1=CC=CC=C1N

Specifications

Color Yellow
Boiling Point 242°C
Quantity 25 g
UN Number 2810
Formula Weight 139.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)aniline