missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)aniline, 98%

Catalog No. aaa1276214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1276214 25 g
AAA1276206 5 g
AAA1276222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1276214 Supplier Thermo Scientific Chemicals Supplier No. A1276214
Only null left

CAS: 2987-53-3 | C7H9NS | 139.216 g/mol

2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water (less).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 2987-53-3
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007708
InChI Key WBRPQQSADOCKCH-UHFFFAOYSA-N
Synonym 2-methylthio aniline, 2-aminothioanisole, 2-methylsulfanyl aniline, 2-methylmercaptoaniline, 2-methylmercapto aniline, o-thioanisidine, o-aminothioanisole, 2-amino thioanisole, benzenamine, 2-methylthio, o-methylthio aniline
PubChem CID 76337
IUPAC Name 2-methylsulfanylaniline
SMILES CSC1=CC=CC=C1N

Specifications

Density 1.111
Boiling Point 233°C to 234°C
Flash Point >110°C (230°F)
Refractive Index 1.624
Quantity 25 g
UN Number UN2810
Beilstein 386211
Solubility Information Soluble in water (less).
Formula Weight 139.23
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)aniline
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 221-055-9
RTECSNumber CY1138333
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only