missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylthio)-2-thiazoline 98.0+%, TCI America™
$67.38 - $331.13
Chemical Identifiers
| CAS | 19975-56-5 |
|---|---|
| Molecular Formula | C4H7NS2 |
| Molecular Weight (g/mol) | 133.23 |
| MDL Number | MFCD00005312 |
| InChI Key | QFGRBBWYHIYNIB-UHFFFAOYSA-N |
| Synonym | 2-(Methylmercapto)-2-thiazoline |
| PubChem CID | 88324 |
| IUPAC Name | 2-(methylsulfanyl)-4,5-dihydro-1,3-thiazole |
| SMILES | CSC1=NCCS1 |
Chemical Identifiers
| 19975-56-5 | |
| 133.23 | |
| QFGRBBWYHIYNIB-UHFFFAOYSA-N | |
| 88324 | |
| CSC1=NCCS1 |
| C4H7NS2 | |
| MFCD00005312 | |
| 2-(Methylmercapto)-2-thiazoline | |
| 2-(methylsulfanyl)-4,5-dihydro-1,3-thiazole |
Specifications
| 19975-56-5 | |
| 217°C | |
| MFCD00005312 | |
| 2-(Methylmercapto)-2-thiazoline | |
| CSC1=NCCS1 | |
| 133.23 | |
| 133.23 | |
| Liquid |
| Yellow | |
| C4H7NS2 | |
| 5 g | |
| QFGRBBWYHIYNIB-UHFFFAOYSA-N | |
| 2-(methylsulfanyl)-4,5-dihydro-1,3-thiazole | |
| 88324 | |
| ≥98.0% (GC) | |
| 2-(Methylthio)-2-thiazoline |
Safety and Handling
TSCA : No