accessibility menu, dialog, popup

UserName

H3CSNS

CAS RN 19975-56-5

IUPAC Name: 2-(methylsulfanyl)-4,5-dihydro-1,3-thiazole
Synonyms: 2-(methylsulfanyl)-4,5-dihydro-1,3-thiazole
Molecular Weight (g/mol): 133.23
Molecular Formula: C4H7NS2
InChi Key: QFGRBBWYHIYNIB-UHFFFAOYSA-N
SMILES: CSC1=NCCS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results