missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiazole 98.0+%, TCI America™

$52.19 - $52.19

Chemical Identifiers

CAS 3581-87-1
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00053144
InChI Key VZWOXDYRBDIHMA-UHFFFAOYSA-N
PubChem CID 77129
IUPAC Name 2-methyl-1,3-thiazole
SMILES CC1=NC=CS1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M17555G
View Documents
TCI America
M17555G
5 g
Each for $52.19
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3581-87-1
99.151
VZWOXDYRBDIHMA-UHFFFAOYSA-N
2-methyl-1,3-thiazole
C4H5NS
MFCD00053144
77129
CC1=NC=CS1

Specifications

3581-87-1
Yellow
C4H5NS
5 g
VZWOXDYRBDIHMA-UHFFFAOYSA-N
2-methyl-1,3-thiazole
77129
≥98.0% (GC)
2-Methylthiazole
-24°C
129°C
MFCD00053144
1993
CC1=NC=CS1
99.151
99.15
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'