accessibility menu, dialog, popup

UserName

CH3SN

CAS RN 3581-87-1

IUPAC Name: 2-methyl-1,3-thiazole
Synonyms: 2-methyl-thiazole
Molecular Weight (g/mol): 99.15
Molecular Formula: C4H5NS
InChi Key: VZWOXDYRBDIHMA-UHFFFAOYSA-N
SMILES: CC1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results