missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiazole 98.0+%, TCI America™

Catalog No. M17555G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17555G Supplier TCI America Supplier No. M17555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3581-87-1
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.151
MDL Number MFCD00053144
InChI Key VZWOXDYRBDIHMA-UHFFFAOYSA-N
PubChem CID 77129
IUPAC Name 2-methyl-1,3-thiazole
SMILES CC1=NC=CS1

Specifications

Melting Point -24°C
Color Yellow
Boiling Point 129°C
Quantity 5 g
UN Number 1993
Formula Weight 99.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylthiazole