missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. M28021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28021G 1 g
M28025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28021G Supplier TCI America Supplier No. M28021G

Chemical Identifiers

CAS 35272-15-2
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD03407332
InChI Key ZHDRDZMTEOIWSX-UHFFFAOYSA-N
Synonym 2-methylthiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylic acid, 4-thiazolecarboxylic acid, 2-methyl, 2-methyl-thiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylicacid, 2-methyl-thiazole4-carboxylic acid, 4-thiazolecarboxylicacid, 2-methyl, 2-methyl-1,3-thiazole-4-carboxylicacid
PubChem CID 284728
IUPAC Name 2-methyl-1,3-thiazole-4-carboxylic acid
SMILES CC1=NC(=CS1)C(=O)O

Specifications

Melting Point 144°C
Quantity 1 g
Formula Weight 143.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylthiazole-4-carboxylic Acid