accessibility menu, dialog, popup

UserName

CAS RN 35272-15-2

IUPAC Name: 2-methyl-1,3-thiazole-4-carboxylic acid
Synonyms: 2-methyl-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 143.16
Molecular Formula: C5H5NO2S
InChi Key: ZHDRDZMTEOIWSX-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)