missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyltetrahydrothiophene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 132°C |
| Quantity | 25 mL |
| UN Number | 1993 |
| Formula Weight | 102.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Methyltetrahydrothiophene |
Safety and Handling
| TSCA | No |