missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyltetrahydrofuran (stabilized with BHT) 98.0+%, TCI America™
Supplier: TCI America M0437100ML
Specifications
| 2-Methyltetrahydrofuran (stabilized with BHT) | |
| Colorless | |
| 100 mL | |
| C5H10O | |
| 2536 | |
| JWUJQDFVADABEY-UHFFFAOYSA-N | |
| 2-methyloxolane | |
| 7301 | |
| ≥98.0% (GC) |
| 96-47-9 | |
| 78°C | |
| Clear Liquid at 20°C | |
| MFCD00005367 | |
| 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran | |
| CC1CCCO1 | |
| 86.134 | |
| 86.13 |
Chemical Identifiers
| 96-47-9 | |
| 86.134 | |
| JWUJQDFVADABEY-UHFFFAOYSA-N | |
| 7301 | |
| CC1CCCO1 |
| C5H10O | |
| MFCD00005367 | |
| 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran | |
| 2-methyloxolane |
Safety and Handling
EINECSNumber : (5)-6944
RTECSNumber : LU2800000
TSCA : Yes