missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran (stabilized with BHT) 98.0+%, TCI America™

Catalog No. p-7132169
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M043725ML 25 mL
M0437100ML 100 mL
M0437500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M043725ML Supplier TCI America Supplier No. M043725ML
Only null left

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00005367
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Color Colorless
Boiling Point 78°C
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 2536
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Methyltetrahydrofuran (stabilized with BHT)
EINECSNumber (5)-6944
RTECSNumber LU2800000
TSCA Yes