missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 98.0+%, TCI America™
$132.35 - $132.35
Chemical Identifiers
| CAS | 57124-87-5 |
|---|---|
| Molecular Formula | C5H10OS |
| Molecular Weight (g/mol) | 118.194 |
| MDL Number | MFCD01632596 |
| InChI Key | DBPHPBLAKVZXOY-UHFFFAOYSA-N |
| Synonym | 2-methyl-3-tetrahydrofuranthiol, 2-methyltetrahydrofuran-3-thiol, tetrahydro-2-methyl-3-furanthiol, 3-furanthiol, tetrahydro-2-methyl, 2-methyl-3-mercaptotetrahydrofuran, pentitol, 1,4-anhydro-2,5-dideoxy-3-thio, 3-mercapto-2-methyltetrahydrofuran, pubchem9972, ksc201q4p, 2-methyl-3-tetrahydrofuran thiol |
| PubChem CID | 62128 |
| IUPAC Name | 2-methyloxolane-3-thiol |
| SMILES | CC1C(CCO1)S |
Chemical Identifiers
| 57124-87-5 | |
| 118.194 | |
| DBPHPBLAKVZXOY-UHFFFAOYSA-N | |
| 62128 | |
| CC1C(CCO1)S |
| C5H10OS | |
| MFCD01632596 | |
| 2-methyl-3-tetrahydrofuranthiol, 2-methyltetrahydrofuran-3-thiol, tetrahydro-2-methyl-3-furanthiol, 3-furanthiol, tetrahydro-2-methyl, 2-methyl-3-mercaptotetrahydrofuran, pentitol, 1,4-anhydro-2,5-dideoxy-3-thio, 3-mercapto-2-methyltetrahydrofuran, pubchem9972, ksc201q4p, 2-methyl-3-tetrahydrofuran thiol | |
| 2-methyloxolane-3-thiol |
Specifications
| 57124-87-5 | |
| 168°C | |
| Clear Liquid at 20°C | |
| MFCD01632596 | |
| 2-methyl-3-tetrahydrofuranthiol, 2-methyltetrahydrofuran-3-thiol, tetrahydro-2-methyl-3-furanthiol, 3-furanthiol, tetrahydro-2-methyl, 2-methyl-3-mercaptotetrahydrofuran, pentitol, 1,4-anhydro-2,5-dideoxy-3-thio, 3-mercapto-2-methyltetrahydrofuran, pubchem9972, ksc201q4p, 2-methyl-3-tetrahydrofuran thiol | |
| CC1C(CCO1)S | |
| 118.194 | |
| 118.19 | |
| 2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) |
| Yellow | |
| 5 g | |
| C5H10OS | |
| 3336 | |
| DBPHPBLAKVZXOY-UHFFFAOYSA-N | |
| 2-methyloxolane-3-thiol | |
| 62128 | |
| ≥98.0% (GC) |
Safety and Handling
TSCA : Yes