accessibility menu, dialog, popup

UserName

CH3OHS

CAS RN 57124-87-5

IUPAC Name: 2-methyloxolane-3-thiol
Synonyms: 2-methyloxolane-3-thiol
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: DBPHPBLAKVZXOY-UHFFFAOYNA-N
SMILES: CC1OCCC1S
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)