Learn More
2-(Methylsulfonyl)acetonitrile, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 2274-42-2 |
|---|---|
| Molecular Formula | C3H5NO2S |
| Molecular Weight (g/mol) | 119.14 |
| InChI Key | FOTRKCAZUSJCQD-UHFFFAOYSA-N |
| Synonym | methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone |
| PubChem CID | 75283 |
| ChEBI | CHEBI:32444 |
| IUPAC Name | 2-methylsulfonylacetonitrile |
| SMILES | CS(=O)(=O)CC#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC438420050
|
Thermo Scientific Chemicals
438420050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC438420250
|
Thermo Scientific Chemicals
438420250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2274-42-2 | |
| 119.14 | |
| methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone | |
| CHEBI:32444 | |
| CS(=O)(=O)CC#N |
| C3H5NO2S | |
| FOTRKCAZUSJCQD-UHFFFAOYSA-N | |
| 75283 | |
| 2-methylsulfonylacetonitrile |
Specifications
| 2274-42-2 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 5g | |
| FOTRKCAZUSJCQD-UHFFFAOYSA-N | |
| 2-methylsulfonylacetonitrile | |
| 75283 | |
| 119.14 | |
| 2-(Methylsulfonyl)acetonitrile |
| 96.0 | |
| 29.03 to 31.45% C, 11.29 to 12.23% N | |
| 97% | |
| C3H5NO2S | |
| methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone | |
| CS(=O)(=O)CC#N | |
| 119.14 | |
| CHEBI:32444 | |
| 97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 218-888-5
RUO â Research Use Only