missing translation for 'onlineSavingsMsg'
Learn More

2-Methylresorcinol 98.0+%, TCI America™

Catalog No. p-7132546
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M110925G 25 g
M1109500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M110925G Supplier TCI America Supplier No. M110925G
Only null left

Chemical Identifiers

CAS 608-25-3
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002271
InChI Key ZTMADXFOCUXMJE-UHFFFAOYSA-N
Synonym 2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
PubChem CID 11843
IUPAC Name 2-methylbenzene-1,3-diol
SMILES CC1=C(C=CC=C1O)O

Specifications

Melting Point 122°C
Quantity 25 g
UN Number 2811
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylresorcinol
EINECSNumber (3)-2766&(4)-0057
RTECSNumber VH2009500
TSCA Yes