missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylquinoxaline 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 245°C |
| Quantity | 5 g |
| Formula Weight | 144.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Methylquinoxaline |
Safety and Handling
| RTECSNumber | VD3450000 |
| TSCA | No |