missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylquinoline 98.0+%, TCI America™
$44.73 - $231.14
Chemical Identifiers
| CAS | 91-63-4 |
|---|---|
| Molecular Formula | C10H9N |
| Molecular Weight (g/mol) | 143.19 |
| MDL Number | MFCD00006756 |
| InChI Key | SMUQFGGVLNAIOZ-UHFFFAOYSA-N |
| Synonym | quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech |
| PubChem CID | 7060 |
| IUPAC Name | 2-methylquinoline |
| SMILES | CC1=CC=C2C=CC=CC2=N1 |
Chemical Identifiers
| 91-63-4 | |
| 143.19 | |
| SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
| 7060 | |
| CC1=CC=C2C=CC=CC2=N1 |
| C10H9N | |
| MFCD00006756 | |
| quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
| 2-methylquinoline |
Specifications
| 91-63-4 | |
| Yellow | |
| C10H9N | |
| 25 g | |
| SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
| 2-methylquinoline | |
| 7060 | |
| ≥98.0% (GC,T) | |
| 2-Methylquinoline |
| -2°C | |
| 247°C | |
| MFCD00006756 | |
| quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
| CC1=CC=C2C=CC=CC2=N1 | |
| 143.19 | |
| 143.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-0795&(5)-0796
RTECSNumber : UZ9625000
TSCA : Yes